Få mere at vide
Ampicillin-d5 (Mixture of Diastereomers), TRC
Artikelnummer. 30334661
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30334661 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334661 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6343021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ampicillin phenyl-d5 (mixture of diastereomers)
CAS 1426173-65-0
Molekylær formel C16 D5 H14 N3 O4 S
Mængde 1 mg
Synonym AY 6108-d5, Albipen-d5, Amfipen-d5, Amplital-d5, Penicline-d5
InChI formel InChI=1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23)/t9?,10-,11+,14-/m1/s1/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(C(N)C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)O)c([2H])c1[2H]
IUPAC navn (2S,5R,6R)-6-[[2-amino-2-(2,3,4,5,6-pentadeuteriophenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvægt (g/mol) 354.44
Formel vægt 354.141