Få mere at vide
Amoxicillin Diketopiperazine, TRC
Artikelnummer. 30006873
Skift visning
Mængde:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006873 50 mg
17900795 5 mg
17912915 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006873 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63423450MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 94659-47-9
Molekylær formel C16 H19 N3 O5 S
Molekylvægt (g/mol) 365.4
Synonym (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Amoxicillin Trihydrate Imp. C (EP), Amoxicillin Diketopiperazines
IUPAC navn (4S)-2-[5-(4-hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
SMIL CC1(C)SC(N[C@H]1C(=O)O)C2NC(=O)C(NC2=O)c3ccc(O)cc3
Tekniske data
Kemisk navn eller materiale (4S)-2-[5-(4-Hydroxyphenyl)-3,6-dioxopiperazin-2-yl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Amoxicillin Diketopiperazines)
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-14(25-16)10-13(22)17-9(12(21)18-10)7-3-5-8(20)6-4-7/h3-6,9-11,14,19-20H,1-2H3,(H,17,22)(H,18,21)(H,23,24)/t9?,10?,11-,14?/m0/s1
Formel vægt 365.1045
Procent renhed >95%