Få mere at vide
Amodiaquine-d10, TRC
Artikelnummer. 30347051
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30347051 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347051 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6342021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amodiaquin-d10
CAS 1189449-70-4
Molekylær formel C20 2H10 H12 Cl N3 O
Mængde 1 mg
Synonym Amodiaquine D10
InChI formel InChI=1S/C20H22ClN3O/c1-3-24(4-2)13-14-11-16(6-8-20(14)25)23-18-9-10-22-19-12-15(21)5-7-17(18)19/h5-12,25H,3-4,13H2,1-2H3,(H,22,23)/i1D3,2D3,3D2,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N(Cc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 2-[[bis(1,1,2,2,2-pentadeuterioethyl)amino]methyl]-4-[(7-chloroquinolin-4-yl)amino]phenol
Molekylvægt (g/mol) 365.92
Formel vægt 365.2079