missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate solution 1mol/L, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Concentration | 1M (1N) |
CAS | 7732-18-5 |
Packaging | Bottle |
Quantity | 1000 mL |
Chemical Name or Material | Ammonium Thiocyanate |
Safety and Handling
Product Identifier
- Ammonium thiocyanate 1M Volumetric Solution
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'