missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Monobasic, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | NH4H2PO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 115.03g/mol |
Percent Purity | ≥99% (T) |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Ammonium Phosphate Monobasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'