missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog Number p-7071742
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
N.OP(O)(O)=O
This item is not returnable. View return policy
7722-76-1
H6NO4P
115.03
MFCD00003396
LFVGISIMTYGQHF-UHFFFAOYSA-N
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
24402
CHEBI:62982
azanium;dihydrogen phosphate
N.OP(O)(O)=O

Puriss. p.a., ACS reagent,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 50 g
Linear Formula NH4H2PO4
UN Number NONH for all modes of transport
Formula Weight 115.03g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Chemical Name or Material Ammonium Phosphate Monobasic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'