missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Dibasic, puriss., Honeywell Fluka™
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 kg |
Linear Formula | (NH4)2HPO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 132.06g/mol |
Percent Purity | 99 to 102% |
Grade | puriss. |
Chemical Name or Material | Ammonium Phosphate Dibasic, Meets analytical specification of NF |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'