missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, ≥98.0%, ACS Reagent, Honeywell™ Fluka™

Catalog Number 15642700
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
4 x 2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
This item is not returnable. View return policy
7727-54-0
H8N2O8S2
228.19
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 120°C
Color Colorless
Physical Form Solid
Quantity 100 g
Linear Formula (NH4)2S2O8
UN Number UN1444
IUPAC Name diazanium;sulfonatooxy sulfate
Formula Weight 228.2g/mol
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Poly Bottle
Residue after Ignition ≤0.05%
Density 1.98g/cm3; Vapor Density: 7.9 (vs air)
Chemical Name or Material Ammonium Persulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium persulfate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'