missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium perrhenate(VII), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 2 g |
Assay Percent Range | 99.999% (Metal basis) |
Solubility Information | Soluble in water. |
IUPAC Name | azanium;oxido(trioxo)rhenium |
Formula Weight | 268.24 |
Appearance | White crystalline |
Density | 3.97 g/cm3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'