Få mere at vide
Ammonium Nonanoate, TRC
Artikelnummer. 30330380
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30330380 5 g
1 options
Artikelnummer. 30330380 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6336955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nonanoic Acid Ammonium
CAS 63718-65-0
Molekylær formel C9 H17 O2 . H4 N
Mængde 5 g
Synonym Ammonium Nonanoate, Nonanoic acid, ammonium salt (9CI), Ammonium nonanoate, Ammonium pelargonate, Pelargonic acid ammonium salt, Nonanoic Acid Ammonium
InChI formel InChI=1S/C9H18O2.H3N/c1-2-3-4-5-6-7-8-9(10)11;/h2-8H2,1H3,(H,10,11);1H3
SMIL O=C(O)CCCCCCCC.N
IUPAC navn ammonia;nonanoic acid
Molekylvægt (g/mol) 175.268
Formel vægt 175.157