missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ammonium (NH4+) Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™
Shop alle Honeywell Fluka produkter
Klik for at se tilgængelige muligheder
Mængde:
100 ml
Beskrivelse
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ammonium Standard for Ion Chromatography |
| Tæthed | 1.01 g/cm3 |
| Mængde | 100 ml |
| Grad | Standards for Ion Chromatography |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion