missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
250 g
500 g
Description
Ammonium Molybdate Tetrahydrate, Extra Pure, SLR
Specifications
Melting Point | 190°C |
Color | White |
pH | 5.0 to 5.5 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 1235.86g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Ammonium molybdate(VI) tetrahydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'