missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(Iii) Sulfate Dodecahydrate, 99%, ACS Reagent, Honeywell™ Fluka™
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 2.5 kg |
Linear Formula | NH4Fe(SO4)2 · 12H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 482.19g/mol |
Percent Purity | 99% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Chemical Name or Material | Ammonium Iron(Iii) Sulfate Dodecahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'