missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium iron(III) oxalate trihydrate, 98%, Thermo Scientific Chemicals
160.47 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
Quantity | 100 g |
UN Number | UN3288 |
Merck Index | 14,517 |
Solubility Information | Soluble in water. Insoluble in ethanol. |
Formula Weight | 428.07 (374.02 Anhydrous) |
Percent Purity | 98% |
Sensitivity | Hygroscopic; Light sensitive |
Density | 1.78 |
Chemical Name or Material | Ammonium iron(III) oxalate trihydrate |
Safety and Handling
Product Identifier
- Ammonium iron(III) oxalate trihydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P321-Specific treatment (see on this label).
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'