missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
2.5 L
6 x 500 mL
12 x 100 mL
Unit Size:
1 L
100mL
2.50 L
25mL
500mL
Pack of 12
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Linear Formula | NH4OH |
Concentration | 28 to 30% NH3 basis |
Beilstein | 3587154 |
Density | 0.9g/mL at 25°C |
Quantity | 6 x 500 mL |
IUPAC Name | azanium;hydroxide |
Explosion Limit | 0.27 |
Formula Weight | 35.05g/mol |
Grade | ACS Reagent |
Packaging | Glass Bottle |
Show More |
Safety and Handling



Product Identifier
- Ammonium hydroxide solution
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ammonia, aqueous solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction