Få mere at vide
Ammonium Formate-d5, CDN
Artikelnummer. 30313795
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313795 1 g -
30310521 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313795 Leverandør CDN Leverandørnr. CDND63431G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ammonium Formate-D5
CAS 65387-23-7
Molekylær formel C D O2 . D4 N
Mængde 1 g
Synonym Formic-d acid-d, ammonium-d3 salt (9 CI), Ammonium Formate-d5, Ammonium Formate-D5, Ammonium-d4 Formate-d, Formic-d acid ammonium-d4 salt
InChI formel InChI=1 S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3/i1D;/hD4
SMIL [2 H]C(=O)[O-].[2 H][N+]([2 H])([2 H])[2 H]
IUPAC navn deuterioformate;tetradeuterioazanium
Molekylvægt (g/mol) 68.0867
Formel vægt 68.0634 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity