Learn More
Ammonium dihydrogen phosphate, Matrix Modifier Solution, Specpure™, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 042663.AE
1187.41 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : CAS Number : 7722-76-1 Weight : 0.10000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium dihydrogen phosphate Matrix Modifier Solution | |
100 mL | |
UN3264 | |
Liquid |
10% NH4H2PO4 in 2% HNO3 | |
MFCD00003396 | |
Specpure™ |
RUO – Research Use Only