Få mere at vide
STEMCELL Technologies Ammonium Chloride Solution
Artikelnummer. 17159311
Encompass_Preferred
Skift visning
Mængde:
100 mL
500 mL
Pakningsstørrelse:
100ml
500ml
Artikelnummer. Mængde unitSize
17159311 100 mL 100ml
15570226 500 mL 500ml
2 options
Artikelnummer. 17159311
Brand: STEMCELL Technologies 07800
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Reagent for lysis of red blood cells
  • Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells
  • It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes
  • It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis
Tekniske data
Mængde 100 mL
Til brug med (applikation) Lysis of Red Blood Cells