Få mere at vide
Ammonium Chloride-d4, TRC
Artikelnummer. 30339108
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30339108 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339108 Leverandør Toronto Research Chemicals Leverandørnr. TRCA633952500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ammonium Chloride-d4
CAS 12015-14-4
Molekylær formel Cl . D4 N
Mængde 500 mg
Synonym Ammonium chloride ((ND4)Cl) (7CI,8CI,9CI), Ammonium chloride-d4, Ammonium-d4 chloride, Deuterated ammonium chloride (ND4Cl), Hydrochloric acid-d, mono(ammoniate-d3), Tetradeuteroammonium chloride, Ammonium chloride-d4
InChI formel InChI=1S/ClH.H3N/h1H;1H3/i/hD4
SMIL [Cl-].[2H][N+]([2H])([2H])[2H]
IUPAC navn tetradeuterioazanium;chloride
Molekylvægt (g/mol) 57.516
Formel vægt 57.028