missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, BAKER ANALYZED™ ACS, J.T. Baker™

Catalog Number p-7220847
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
N.Cl
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
N.Cl

Meets ACS Specifications. Meets Reagent Specificatons for testing USP/NF monographs.

Melting Point 335°C
pH 4.5 to 5.5 (5% Solution at 25°C)
Quantity 500 g
Assay Percent Range min. 99.5%
IUPAC Name amine hydrochloride
Packaging Plastic bottle for solids
Residue after Ignition max. 0.01%
Density 1.5256 g/cm³ (20°C)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'