missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, ACS Reagent, Honeywell Fluka™

Catalog Number p-7071872
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
4 x 2.5 kg
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206
This item is not returnable. View return policy
12125-02-9
ClH4N
53.49
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
25517
CHEBI:31206

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 25 g
Linear Formula NH4Cl
Beilstein 4371014
SMILES N.Cl
IUPAC Name azanium;chloride
Formula Weight 53.49g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Residue after Ignition ≤0.01%
Vapor Pressure 1mmHg (160.4°C)
Density Vapor Density: 1.9 (vs air)
Chemical Name or Material Ammonium Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'