missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | NH4Cl |
Beilstein | 4371014 |
SMILES | N.Cl |
IUPAC Name | azanium;chloride |
Formula Weight | 53.49g/mol |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Residue after Ignition | ≤0.01% |
Vapor Pressure | 1mmHg (160.4°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'