Learn More
Ammonium cerium(IV) Sulfate Dihydrate, Supelco™
Product Code. 15807090
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity Unit Size
15807090 100 g 100g
1 options
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
Product Code. 15807090 Supplier Supelco Supplier No. 1022730100
 Restricted Item
Forventet forsendelse: 18-11-2026
MQ300 Graded - For Products Used in Applications Requiring Enhanced Change Control and Quality Agreement
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Specifications
CAS 10378-47-9
Quantity 100 g
Molecular Formula (NH4)4Ce(SO4)4 · 2H2O
MDL Number MFCD00149417
Synonym Ammonium cerium(IV) sulfate dihydrate, Cerium(IV) ammonium sulfate, Ammonium cerium(IV) sulfate dihydrate
Molecular Weight (g/mol) 632.55
Grade Research
Chemical Name or Material Ammonium Cerium(IV) Sulfate Dihydrate