missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Nitrate, ACS Reagent, ≥98.5%, Honeywell Fluka™

Catalog Number p-7120169
Click to view available options
Quantity:
50 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
16774-21-3
MFCD00151121
This item is not returnable. View return policy
16774-21-3
MFCD00151121

ACS Reagent,≥98.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 214°C
Color Orange
pH Acidic
Quantity 50 g
Linear Formula Ce(NH4)2(NO3)6
UN Number UN3085
Formula Weight 548.22g/mol
Percent Purity ≥98.5%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder/Crystals
Density 2.49g/cu.cm
Chemical Name or Material Ammonium Cerium(Iv) Nitrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'