missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3776841
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
HDPE plastic bottle
506-87-6
CH8N2O3
96.09
MFCD00010890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]
506-87-6
CH8N2O3
96.09
MFCD00010890
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
517111
diazanium;carbonate
C(=O)([O-])[O-].[NH4+].[NH4+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58.0°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Linear Formula (NH4)2CO3
Merck Index 15, 505
Solubility Information Solubility in water: soluble (320g/L at 20°C)
Formula Weight 96.09
Percent Purity ≥30.0% (NH3)
Grade ACS Reagent
Packaging HDPE plastic bottle
Chemical Name or Material Ammonium carbonate, For analysis ACS
Show More Show Less
Product Identifier
  • Ammonium carbonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'