missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, Extra Pure, SLR, Crystals, Fisher Chemical™
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Specifications
Melting Point | 114°C |
Color | White |
pH | 7 |
Packaging | Plastic powder jar |
Molecular Formula | C2H7NO2 |
Quantity | 500 g |
Formula Weight | 77.08g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'