missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, Extra Pure, SLR, Crystals, Fisher Chemical™

Catalog Number 10468010
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
631-61-8
77.08
13066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O
This item is not returnable. View return policy
631-61-8
77.08
13066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate
[NH4+].CC([O-])=O
Melting Point 114°C
Color White
pH 7
Packaging Plastic powder jar
Molecular Formula C2H7NO2
Quantity 500 g
Formula Weight 77.08g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'