missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 25 g |
Linear Formula | CH3CO2NH4 |
Beilstein | 4186741 |
Formula Weight | 77.08g/mol |
Percent Purity | ≥97% |
Grade | ACS Reagent |
Residue after Ignition | ≤0.01% |
Chemical Name or Material | Ammonium Acetate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'