missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, ACS Reagent, Honeywell Fluka™

Product Code. 15632380
Click to view available options
Quantity:
25 g
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]

Specifications

Quantity 25 g
Linear Formula CH3CO2NH4
Beilstein 4186741
Formula Weight 77.08g/mol
Percent Purity ≥97%
Grade ACS Reagent
Residue after Ignition ≤0.01%
Chemical Name or Material Ammonium Acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'