missing translation for 'onlineSavingsMsg'
Få mere at vide

Ammonium-15N Bromide, TRC

Artikelnummer. 30345655
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345655 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30345655 Leverandør Toronto Research Chemicals Leverandørnr. TRCA633847100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Ammonium-15N Bromide
CAS 39466-31-4
Molekylær formel BrH415N
Mængde 100 mg
InChI formel InChI=1S/BrH.H3N/h1H;1H3/i;1+1
SMIL [15NH3].Br
IUPAC navn azane;hydrobromide
Molekylvægt (g/mol) 98.94
Formel vægt 97.9497
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.