missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Amemid, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | Ammelide |
| CAS | 645-93-2 |
| Analyt- eller komponentnavne | Ammelide |
| Forsendelsestilstand | Stuetemperatur |
| Molekylær formel | C3 H4 N4 O2 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Ammelide (6CI), s-Triazine-2,4(1H,3H)-dione, 6-amino- (8CI), s-Triazine-2,4(1H,3H)-dione, dihydro-6-imino- (7CI), 6-Amino-1,3,5-triazine-2,4(1H,3H)-dione, 2-Amino-1,3,5-triazine-4,6-dione, 2-Amino-4,6-dihydroxy-s-triazine, Ammelid, GS 35713, Melanurenic acid, NSC 8146, s-Dihydroxyaminotriazine |
| InChI formel | InChI=1S/C3H4N4O2/c4-1-5-2(8)7-3(9)6-1/h(H4,4,5,6,7,8,9) |
| Løsningstype | Fedt |
| SMIL | Nc1nc(O)nc(O)n1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?