Få mere at vide
Amlodipine N-Lactoside (Mixture of Diastereomers), TRC
Artikelnummer. 17955005
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17955005 10 mg
17924195 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17955005 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63352010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2173291-00-2
Molekylær formel C32 H45 Cl N2 O15
Molekylvægt (g/mol) 733.16
Synonym Amlodipine N-Lactoside (Mixture of Diastereomers)
IUPAC navn O3-ethyl O5-methyl 4-(2-chlorophenyl)-2-[2-[[(3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydropyran-2-yl]oxy-tetrahydropyran-2-yl]amino]ethoxymethyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
SMIL C(OCC)(=O)C=1C(C(C(OC)=O)=C(C)NC1COCCNC2O[C@H](CO)[C@@H](O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O)C4=C(Cl)C=CC=C4
Tekniske data
Kemisk navn eller materiale Amlodipine N-Lactoside (mixture of diastereomers)
Analyt- eller komponentnavne Amlodipine N-Lactoside (mixture of diastereomers)
Forsendelsestilstand Room Temperature
InChI formel InChi=1S/C32H45ClN2O15/c1-4-47-31(44)22-17(35-14(2)20(30(43)45-3)21(22)15-7-5-6-8-16(15)33)13-46-10-9-34-29-26(41)25(40)28(19(12-37)48-29)50-32-27(42)24(39)23(38)18(11-36)49-32/h5-8,18-19,21,23-29,32,34-42H,4,9-13H2,1-3H3/t18-,19-,21?,23+,24+,25-,26-,27-,28-,29?,32+/m1/s1
Løsningstype Neat
Industry Type Forensics
Mængde 10 mg
Formel vægt 732.25