missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Amlodipine Maleate Adduct, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Amlodipine aspartate |
| CAS | 400602-35-9 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C24 H29 Cl N2 O9 |
| Mængde | 50 mg |
| Synonym | 4-(2-Chlorophenyl)-2-[[2-[(1,2-dicarboxyethyl)amino]ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-Pyridinedicarboxylic Acid 3-Ethyl 5-Methyl Ester, Amlodipine aspartate, L-Aspartate amlodipine, Amlodipine Aspartic Acid Impurity |
| InChI formel | InChI=1S/C24H29ClN2O9/c1-4-36-24(33)21-17(12-35-10-9-26-16(22(30)31)11-18(28)29)27-13(2)19(23(32)34-3)20(21)14-7-5-6-8-15(14)25/h5-8,16,20,26-27H,4,9-12H2,1-3H3,(H,28,29)(H,30,31) |
| SMIL | O=C(OC)C1=C(NC(=C(C(=O)OCC)C1C=2C=CC=CC2Cl)COCCNC(C(=O)O)CC(=O)O)C |
| IUPAC navn | 2-[2-[[4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl-6-methyl-1,4-dihydropyridin-2-yl]methoxy]ethylamino]butanedioic acid |
| Molekylvægt (g/mol) | 524.95 |
| Vis mere |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?