Learn More
Amlodipine EP Impurity B, Mikromol™
Product Code. 30460453
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460453 4 x 25 mg 100mg
17960676 25 mg 25mg
17900866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30460453 Supplier Mikromol Supplier No. MM0383.0200254AMP
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Amlodipine EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[(2-[(2-(methylcarbamoyl)benzoyl)amino]ethoxy)methyl]-1,4-dihydropyridine-3,5-dicarboxylate
CAS 721958-72-1
Analyte or Component Names 3-ethyl 5-methyl (4RS)-4-(2-chlorophenyl)-6-methyl-2-[(2-[(2-(methylcarbamoyl)benzoyl)amino]ethoxy)methyl]-1,4-dihydropyridine-3,5-dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C29 H32 Cl N3 O7
Certifications/Compliance ISO 17025
Synonym 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-6-methyl-2-[[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. B (EP), Amlodipine Imp. B (EP), 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-6-methyl-2-[[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxy]methyl]-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Related Compound B, Amlodipine Besilate Impurity B, Amlodipine Impurity B
InChI Formula InChI=1S/C29H32ClN3O7/c1-5-40-29(37)25-22(16-39-15-14-32-27(35)19-11-7-6-10-18(19)26(34)31-3)33-17(2)23(28(36)38-4)24(25)20-12-8-9-13-21(20)30/h6-13,24,33H,5,14-16H2,1-4H3,(H,31,34)(H,32,35)
Solution Type Neat
SMILES CCOC(=O)C1=C(COCCNC(=O)c2ccccc2C(=O)NC)NC(=C(C1c3ccccc3Cl)C(=O)OC)C
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2-chlorophenyl)-2-methyl-6-[2-[[2-(methylcarbamoyl)benzoyl]amino]ethoxymethyl]-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 570.03
Quantity 4 x 25 mg
Formula Weight 569.1929 g/mol