Få mere at vide
Amlodipine EP Impurity A (Amlodipine USP Related Compound D), Mikromol™
Artikelnummer. 30050639
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050639 100 mg
30050349 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050639 Leverandør Mikromol Leverandørnr. MM0383.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Amlodipine EP Impurity A (Amlodipine USP Related Compound D)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-([2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 88150-62-3
Analyt- eller komponentnavne 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-([2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Forsendelsestilstand Room Temperature
Molekylær formel C28 H27 Cl N2 O7
Certificeringer/compliance ISO 17025
Synonym 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. A (EP), Amlodipine Imp. A (EP), Amlodipine USP Related Compound D, Amlodipine USP RC D, 3-Ethyl 5-Methyl (4RS)-4-(2-Chlorophenyl)-2-[[2-(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Phthaloyl Amlodipine, Amlodipine Related Compound D, Amlodipine Besilate Impurity A, Amlodipine Impurity A
InChI formel InChI=1S/C28H27ClN2O7/c1-4-38-28(35)24-21(15-37-14-13-31-25(32)17-9-5-6-10-18(17)26(31)33)30-16(2)22(27(34)36-3)23(24)19-11-7-8-12-20(19)29/h5-12,23,30H,4,13-15H2,1-3H3
SMIL CCOC(=O)C1=C(COCCN2C(=O)c3ccccc3C2=O)NC(=C(C1c4ccccc4Cl)C(=O)OC)C
IUPAC navn 3-O-ethyl 5-O-methyl 4-(2-chlorophenyl)-2-[2-(1,3-dioxoisoindol-2-yl)ethoxymethyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 538.98
Mængde 100 mg
Formel vægt 538.1507 g/mol