Få mere at vide
Amlodipine-d4 Maleic Acid Salt, TRC
Artikelnummer. 30341537
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30341537 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341537 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63350225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amlodipine-d4 Maleic Acid Salt
CAS 1185246-15-4
Renhedsgrad noter HPLC
Molekylær formel C24H25D4ClN2O9
Mængde 25 mg
Synonym 2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl)-6-methyl-1,4-dihydropyridine-d4 Maleic Acid Salt
InChI formel InChI=1S/C20H25ClN2O5.C4H4O4/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;5-3(6)1-2-4(7)8/h5-8,17,23H,4,9-11,22H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/i9D2,10D2;
SMIL NC([2H])([2H])C([2H])([2H])OCC1=C(C(OCC)=O)C(C2=CC=CC=C2Cl)C(C(OC)=O)=C(C)N1.O=C(O)/C=C\C(O)=O
IUPAC navn (Z)-but-2-enedioic acid;3-O-ethyl 5-O-methyl 2-[(2-amino-1,1,2,2-tetradeuterioethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvægt (g/mol) 528.97
Procent renhed >95