Få mere at vide
Amitriptyline-d6 Hydrochloride, TRC
Artikelnummer. 30347723
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30347723 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347723 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6333511MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amitriptyline-d6 Hydrochloride
CAS 203645-63-0
Renhedsgrad noter HPLC
Molekylær formel C20 2H6 H17 N . Cl H
Mængde 1 mg
Synonym 1-Propanamine, 3-(10,11-dihydro-5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-di(methyl-d3)-, hydrochloride (9CI), Amitriptyline-d6 hydrochloride, Amitriptyline-D6 hydrochloride (N,N-dimethyl D6)
InChI formel InChI=1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H/i1D3,2D3;
SMIL Cl.[2H]C([2H])([2H])N(CCC=C1c2ccccc2CCc3ccccc13)C([2H])([2H])[2H]
IUPAC navn 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 319.9
Formel vægt 319.1974
Procent renhed >95