Få mere at vide
Amitriptyline-d6 HCl (N,N-dimethyl-d6), CDN
Artikelnummer. 30317802
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317802 0.05 g
30320500 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317802 Leverandør CDN Leverandørnr. CDND35770.05G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amitriptyline-d6 Hydrochloride
CAS 203645-63-0
Molekylær formel C20 2H6 H17 N . Cl H
Mængde 0.05 g
Synonym 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-di(methyl-d3)-, hydrochloride (9 CI), Amitriptyline-d6 hydrochloride, Amitriptyline-D6 hydrochloride (N,N-dimethyl D6)
InChI formel InChI=1 S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12 H,7,13-15H2,1-2H3;1 H/i1D3,2D3;
SMIL Cl.[2 H]C([2 H])([2 H])N(CCC=C1c2ccccc2CCc3ccccc13)C([2 H])([2 H])[2 H]
IUPAC navn 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7 ]annulen-11-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 319.9
Formel vægt 319.1974 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity