Få mere at vide
Amitriptyline-d3 HCl (N-methyl-d3), CDN
Artikelnummer. 30314190
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314190 0.01 g
30314044 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314190 Leverandør CDN Leverandørnr. CDND18300.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amitriptyline-D3 Hydrochloride
CAS 342611-00-1
Molekylær formel C20 2H3 H20 N . Cl H
Mængde 0.01 g
Synonym 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-N-(methyl-d3)-, hydrochloride (9 CI), Amitriptyline-D3 Hydrochloride, Amitriptyline-D3 hydrochloride (N-methyl D3)
InChI formel InChI=1 S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12 H,7,13-15H2,1-2H3;1 H/i1D3;
SMIL Cl.[2 H]C([2 H])([2 H])N(C)CCC=C1c2ccccc2CCc3ccccc13
IUPAC navn 3-(5,6-dihydrodibenzo[2,1-b:1',2'-e][7 ]annulen-11-ylidene)-N-methyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 316.88
Formel vægt 316.1786 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity