Få mere at vide
Amitraz, MedChemExpress
Artikelnummer. 16468616
Encompass_Preferred
Skift visning
Mængde:
100 mg
Pakningsstørrelse:
100mg
Artikelnummer. Mængde unitSize
16468616 100 mg 100mg
1 options
Artikelnummer. 16468616 Leverandør Medchem Express Leverandørnr. HYB1111
MedChemExpress Amitraz is a non-systemic acaricide and insecticide, with alpha-adrenergic agonist activity, interaction with octopamine receptors of the central nervous system and inhibition of monoamine oxidases and prostaglandin synthesis.
Amitraz is a non-systemic acaricide and insecticide, with alpha-adrenergic agonist activity, interaction with octopamine receptors of the central nervous system and inhibition of monoamine oxidases and prostaglandin synthesis.
Tekniske data
Kemisk navn eller materiale Amitraz
Fysisk form Solid
CAS 33089-61-1
Farve Light Yellow
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C19H23N3
Til brug med (applikation) COVID-19-immunoregulation
Synonym BTS-27419
Opløselighedsinformation DMSO : 100 mg/mL (340.82 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL CN(/C=N/C1=CC=C(C)C=C1C)/C=N/C2=CC=C(C)C=C2C
Molekylvægt (g/mol) 293.41
Formel vægt 293.41
Procent renhed 95.0%