Få mere at vide
Amiprofos-methyl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Amiprofos-methyl |
| CAS | 36001-88-4 |
| Molekylær formel | C11 H17 N2 O4 P S |
| Mængde | 250 mg |
| Synonym | Phosphoramidothioic acid, (1-methylethyl)-, O-methyl O-(4-methyl-2-nitrophenyl) ester (9CI), Amiprofos-methyl, Amiprophos-methyl, BAY-NTN 6867, BAY-NTN 80, NSC 313446, NTN 6867, NTN 80, O-(2-Nitro-4-methylphenyl) O-methyl N-isopropylphosphoramidothioate, O-Methyl O-4-methyl-2-nitrophenyl N-isopropylphosphoramidothioate, Tokunol M |
| InChI formel | InChI=1S/C11H17N2O4PS/c1-8(2)12-18(19,16-4)17-11-6-5-9(3)7-10(11)13(14)15/h5-8H,1-4H3,(H,12,19) |
| SMIL | COP(=S)(NC(C)C)Oc1ccc(C)cc1[N+](=O)[O-] |
| IUPAC navn | N-[methoxy-(4-methyl-2-nitrophenoxy)phosphinothioyl]propan-2-amine |
| Molekylvægt (g/mol) | 304.3 |
| Formel vægt | 304.0647 |