Få mere at vide
Amiphenazole Hydrochloride, LoGiCal
Artikelnummer. 30458274
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458274 10 mg
1 options
Artikelnummer. 30458274 Leverandør LoGiCal Leverandørnr. LGCFOR1398.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Amiphenazole Hydrochloride
CAS 942-31-4
Analyt- eller komponentnavne Amiphenazole Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C9 H9 N3 S . Cl H
Certificeringer/compliance ISO 9001
Synonym 5-Phenyl-2,4-thiazolediamine monohydrochloride, 2,4-Diamino-5-phenyl-thiazole hydrochloride, 2,4-Diamino-5-phenyl-thiazole monohydrochloride, Amiphenazole hydrochloride, Phenamizole hydrochloride
InChI formel InChI=1S/C9H9N3S.ClH/c10-8-7(13-9(11)12-8)6-4-2-1-3-5-6;/h1-5H,10H2,(H2,11,12);1H
Løsningstype Neat
SMIL Cl.Nc1nc(N)c(s1)c2ccccc2
IUPAC navn 5-phenyl-1,3-thiazole-2,4-diamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 227.71
Mængde 10 mg
Formel vægt 227.0284 g/mol