Få mere at vide
Amiodarone EP Impurity C (as Hydrochloride), Mikromol™
Beskrivelse
Amiodarone EP Impurity C (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodo-phenyl]methanone Hydrochloride |
| CAS | 1397201-93-2 |
| Analyt- eller komponentnavne | (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodo-phenyl]methanone Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H30 I N O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone Hydrochloride, Amiodarone Hydrochloride Imp. C (EP), Amiodarone Imp. C (EP), (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]-3-iodo-phenyl]methanone Hydrochloride, Amiodarone Hydrochloride Imp. C (EP) as Hydrochloride, Amiodarone Hydrochloride Impurity C as Hydrochloride, Amiodarone Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C25H30INO3.ClH/c1-4-7-11-23-24(19-10-8-9-12-21(19)30-23)25(28)18-13-14-22(20(26)17-18)29-16-15-27(5-2)6-3;/h8-10,12-14,17H,4-7,11,15-16H2,1-3H3;1H |
| SMIL | Cl.CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)c(I)c3 |
| IUPAC navn | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3-iodophenyl]methanone;hydrochloride |
| Vis mere |