Få mere at vide
Amiodarone EP Impurity A (as Oxalate), Mikromol™
Beskrivelse
Amiodarone EP Impurity A (as Oxalate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]phenyl]methanone Oxalate |
| CAS | 2984442-50-2 |
| Analyt- eller komponentnavne | (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]phenyl]methanone Oxalate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H31 N O3 . C2 H2 O4 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Amiodarone Hydrochloride Imp. A (EP), Amiodarone Imp. A (EP), (2-Butylbenzofuran-3-yl)[4-[2-(diethylamino)ethoxy]phenyl]methanone Oxalate, Amiodarone Hydrochloride Imp. A (EP) as Oxalate, Amiodarone Hydrochloride Impurity A as Oxalate, Amiodarone Impurity A as Oxalate |
| InChI formel | InChI=1S/C25H31NO3.C2H2O4/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;3-1(4)2(5)6/h8-10,12-16H,4-7,11,17-18H2,1-3H3;(H,3,4)(H,5,6) |
| SMIL | CCCCc1oc2ccccc2c1C(=O)c3ccc(OCCN(CC)CC)cc3.OC(=O)C(=O)O |
| IUPAC navn | (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]phenyl]methanone;oxalic acid |
| Vis mere |