Få mere at vide
(Aminocarbonyl)azanyl Carbamic Acid Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (Aminocarbonyl)azanyl Carbamic Acid Ester |
| CAS | 4543-62-8 |
| Molekylær formel | C2 H5 N3 O3 |
| Mængde | 10 mg |
| Synonym | Carbamoyloxyurea, (Aminocarbonyl)azanyl Carbamate, N,O-Dicarbamoyl-hydroxylamine, [(Aminocarbonyl)oxy]-urea, N,O-Bis(aminocarbonyl)-hydroxylamine, N-(Carbamoyloxy)urea |
| InChI formel | InChI=1S/C2H5N3O3/c3-1(6)5-8-2(4)7/h(H2,4,7)(H3,3,5,6) |
| SMIL | NC(=O)NOC(=O)N |
| IUPAC navn | (carbamoylamino) carbamate |
| Molekylvægt (g/mol) | 119.079 |
| Formel vægt | 119.033 |