Få mere at vide
γ-Aminobutyric Acid-d6, TRC
Artikelnummer. 30324117
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30324117 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324117 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6029225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale gamma-Aminobutyric Acid-d6
CAS 70607-85-1
Renhedsgrad noter HPLC
Molekylær formel C4H3D6NO2
Mængde 5 mg
Synonym 4-Aminobutanoic-2,2,3,3,4,4-d6 Acid, 4-Aminobutanoic αcid-d6, 3-Carboxypropylamine-d6, 4-Aminobutyric Acid-d6, Aminalon-d6, GABA-d6, Gaballon-d6, Gamarex-d6, Gammalon-d6, Gammalone-d6, Gammar-d6, Gammasol-d6, Mielogen-d6, Mielomade-d6, NSC 27418-d6, NSC 32044-d6, NSC 45460-d6, NSC 51295-d6, Oryza GABA Extract HC 90-d6, Pharma-GABA 20S-d6, Pharmagaba-d6, Pharmagaba 20-d6, Pharmagaba 20D-d6, Piperidic Acid-d6, Piperidinic Acid-d6, ω-Aminobutyric Acid-d6
InChI formel InChI=1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)/i1D2,2D2,3D2
SMIL C(CC(=O)O)CN
IUPAC navn 4-amino-2,2,3,3,4,4-hexadeuteriobutanoic acid
Molekylvægt (g/mol) 109.16
Formel vægt 109.101
Procent renhed >95