Få mere at vide
Aminoacridine Hydrochloride Monohydrate (9-Aminoacridine Hydrochloride Monohydrate), LoGiCal
Artikelnummer. 30460996
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460996 10 mg
1 options
Artikelnummer. 30460996 Leverandør LoGiCal Leverandørnr. LGCFOR1588.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Aminoacridine hydrochloride monohydrate
CAS 52417-22-8
Analyt- eller komponentnavne 9-Aminoacridine hydrochloride monohydrate
Forsendelsestilstand Room Temperature
Molekylær formel C13 H10 N2 . Cl H . H2 O
Certificeringer/compliance ISO 9001
Synonym 9-Acridinamine, hydrochloride, hydrate, 9-Acridinamine, monohydrochloride, monohydrate;9-Aminoacridine hydrochloride monohydrate
InChI formel InChI=1S/C13H10N2.ClH.H2O/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13;;/h1-8H,(H2,14,15);1H;1H2
Løsningstype Neat
SMIL O.Cl.Nc1c2ccccc2nc3ccccc13
IUPAC navn acridin-9-amine;hydrate;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 248.71
Mængde 10 mg
Formel vægt 248.0716 g/mol