Få mere at vide
Aminitrozole, LoGiCal
Artikelnummer. 30463424
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463424 10 mg
1 options
Artikelnummer. 30463424 Leverandør LoGiCal Leverandørnr. LGCFOR1745.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Acetamido-5-nitrothiazole
CAS 140-40-9
Analyt- eller komponentnavne 2-Acetamido-5-nitrothiazole
Forsendelsestilstand Room Temperature
Molekylær formel C5 H5 N3 O3 S
Certificeringer/compliance ISO 9001
Synonym Acetamide, N-(5-nitro-2-thiazolyl)-Thiazole, 2-acetamido-5-nitro- (6CI,7CI), 2-(Acetylamino)-5-nitrothiazole, 2-Acetamido-5-nitrothiazole, 5-Nitro-2-acetamidothiazole, Acetyl enheptin, Acinitrazol, Acinitrazole, Ametoterina, Aminitrozol, Aminitrozole, CL 5279, Cyzine Premix, Enheptin A, Gynofon, Lavoflagin, N-(5-Nitro-2-thiazolyl)acetamide, NSC 31539, NSC 45914, Nitazol, Nitazole, Nithiamide, Pleocide, Trichlorad, Trichocid, Trichoman, Trichorad, Trichoral, Tricogen, Tricolaval, Tricoral, Tricosteril, Trikolaval, Tritheon
InChI formel InChI=1S/C5H5N3O3S/c1-3(9)7-5-6-2-4(12-5)8(10)11/h2H,1H3,(H,6,7,9)
Løsningstype Neat
SMIL CC(=O)Nc1ncc(s1)[N+](=O)[O-]
IUPAC navn N-(5-nitro-1,3-thiazol-2-yl)acetamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 187.18
Mængde 10 mg
Formel vægt 187.0052 g/mol