missing translation for 'onlineSavingsMsg'
Få mere at vide

Amifostine, MedChemExpress

Artikelnummer. 16498558
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16498558 10 mg
16404437 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16498558 Leverandør Medchem Express Leverandørnr. HYB0639

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Amifostine (WR2721) is a broad-spectrum cytoprotective agent and a radioprotector. Amifostine selectively protects normal tissues from damage caused by radiation and chemotherapy. Amifostine is potent hypoxia-inducible factor-α1 (HIF-α1) and p53 inducer. Amifostine protects cells from damage by scavenging oxygen-derived free radicals. Amifostine reduces renal toxicity and has antiangiogenic action.

Amifostine (WR2721) is a broad-spectrum cytoprotective agent and a radioprotector. Amifostine selectively protects normal tissues from damage caused by radiation and chemotherapy. Amifostine is potent hypoxia-inducible factor-α1 (HIF-α1) and p53 inducer. Amifostine protects cells from damage by scavenging oxygen-derived free radicals. Amifostine reduces renal toxicity and has antiangiogenic action.

Tekniske data

Kemisk navn eller materiale Amifostine
Fysisk form Solid
CAS 20537-88-6
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C5H15N2O3PS
Til brug med (applikation) Cancer-Kinase/protease
Synonym WR2721
Opløselighedsinformation H2O : 100 mg/mL (466.81 mM; Need ultrasonic) ∣DMF : 1 mg/mL (4.67 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMIL OP(SCCNCCCN)(O)=O
Molekylvægt (g/mol) 214.22
Formel vægt 214.22
Procent renhed 98.0%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.