Få mere at vide
Amidephrine Mesilate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Amidephrine Mesilate |
| CAS | 1421-68-7 |
| Analyt- eller komponentnavne | Amidephrine Mesilate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H16 N2 O3 S . C H4 O3 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Methanesulfonamide, N-[3-[1-hydroxy-2-(methylamino)ethyl]phenyl]-, monomethanesulfonate (salt) (9CI), Methanesulfonanilide, 3'-[1-hydroxy-2-(methylamino)ethyl]-, monomethanesulfonate (salt) (8CI), Methanesulfonic acid, compd. with 3'-[1-hydroxy-2-(methylamino)ethyl]methanesulfonanilide (7CI), (±)-Amidephrine mesylate, 3'-[1-Hydroxy-2-(methylamino)ethyl]methanesulfonanilide methanesulfonate, Amidefrine mesilate, Amidefrine mesylate, Amidephrine mesylate, Amidephrine monomethanesulfonate, Dircol, Fentrinol, MJ 5190, Nalde, Product 5190, Methanesulfonamide, N-[3-[1-hydroxy-2-(methylamino)ethyl]phenyl]-, methanesulfonate (1:1) |
| InChI formel | InChI=1S/C10H16N2O3S.CH4O3S/c1-11-7-10(13)8-4-3-5-9(6-8)12-16(2,14)15;1-5(2,3)4/h3-6,10-13H,7H2,1-2H3;1H3,(H,2,3,4) |
| Løsningstype | Neat |
| SMIL | CNCC(O)c1cccc(NS(=O)(=O)C)c1.CS(=O)(=O)O |
| Vis mere |